missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LGSN (aa 98-186) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65252 (PA5-65252. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May act as a component of the cytoskeleton or as a chaperone for the reorganization of intermediate filament proteins during terminal differentiation in the lens. Does not seem to have enzymatic activity.
Specifications
Specifications
| Accession Number | Q5TDP6 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51557 |
| Name | Human LGSN (aa 98-186) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | GLULD1; glutamate-ammonia ligase (glutamine synthase) domain containing 1; glutamate-ammonia ligase domain-containing protein 1; Lengsin; lengsin, lens protein with glutamine synthetase domain; Lens glutamine synthase-like; LGS; LGSN |
| Common Name | LGSN |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction