missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB2/CD85d/ILT4 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
181.00 € - 453.00 €
Specifications
| Antigen | ILT4/CD85d/LILRB2 |
|---|---|
| Clone | 287219 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
ILT4/CD85d/LILRB2 Monoclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Flow Cytometry.Specifications
| ILT4/CD85d/LILRB2 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
| LILRB2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human ILT4/CD85d in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT6, rhILT7, rhILT11, rhLIR6 or rhLIR8 is observed. |
| 287219 | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| Mouse | |
| Human | |
| ACT64556 | |
| 10288 | |
| Mouse myeloma cell line NS0-derived recombinant human ILT4/CD85d Gly24-His458 Accession # ACT64556 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title