missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human LRRC19 (NP_075052) VersaClone cDNA

Product Code. 18724238
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18724238 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18724238 Supplier R&D Systems™ Supplier No. RDC1756

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Leucine-rich Repeat Containing 19

The Human LRRC19 (NP_075052) VersaClone cDNA clone contains the complete ORF for LRRC19, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LRRC19 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Ordreinfo

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Tekniske data

Product Type cDNA
Gene hLRRC19
Accession Number NP_075052
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1126bp
Gene ID (Entrez) 26762 (Human), 171283 (Mouse), 286934 (Rat), 102141332 (Cynomolgus Monkey)
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.