missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRTM2 Synthetic Peptide

Product Code. 15285209
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15285209 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15285209 Supplier Invitrogen™ Supplier No. PEP1211
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 17 amino acid peptide near the carboxy terminus of human LRRTM2. PEP-1211 can be used as a blocking peptide with polyclonal antibody PA5-21097.

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM2 interacts with PSD-95 and regulates the surface expression of AMPA receptors. LRRTM2 also functions as a neurexin ligand, binding both Neurexin 1-alpha and -beta, suggesting that LRRTM2-Neurexin1 interaction plays a critical role in regulatory excitatory synapse development.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 26045
Species Human
Purification Method purified
Accession Number O43300
Common Name LRRTM2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias AI851755; BB129880; C630011A14Rik; KIAA0416; leucine rich repeat transmembrane neuronal 2; leucine-rich repeat neuronal 2 protein; leucine-rich repeat transmembrane neuronal 2 protein; leucine-rich repeat transmembrane neuronal protein 2; LRRN2; LRRTM2; mKIAA0416
Gene Symbol LRRTM2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.