missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Ly6K (NP_059997) VersaClone cDNA
Description
- Also known as: CT97; ly-6K; URLC10; HSJ001348
- Ly6K is a member of the Ly-6 antigen family of molecules.
- It is a GPI-linked glycoprotein.
- It has restricted expression to testis and skin.
- Ly6K is found in/on carcinomas and is known to circulate in normal and tumor-patient blood.
- Alternative spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hLY6K |
| Accession Number | NP_059997 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 511 bp |
| Gene ID (Entrez) | 54742 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?