missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human M-CSF Recombinant Protein

Product Code. 10336479
Change view
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10336479 100 μg Each
10089418 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336479 Supplier Gibco™ Supplier No. PHC9501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Reconstitution: Lyophilized human M-CSF should be reconstituted in 20mM Tris-HCl, pH 8.0 to a concentration of 0.1-1.0 mg/mL. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: Lyophilized human M-CSF should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted human M-CSF at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09603
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 38 kDa
Name Human M-CSF
Purification Method Purified
Quantity 100 μg
Storage Requirements 4°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1
Common Name M-CSF
Gene Symbol Csf1
Biological Activity ED50 ≤ 5 ng/mL; determined by the dose-dependent proliferation of M-NSF60 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage 4°C
Activity ED50 ≤ 5 ng/mL; determined by the dose-dependent proliferation of M-NSF60 cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Wet Ice
Expression System HEK293 cells
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype GM-CSF, G-CSF, & M-CSF
Research Category Cardiovascular Research, Stem Cell Research, Differentiation, Inflammation
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.