missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Magnetic Luminex™ Performance Assay Base Kit A

Product Code. 30564783
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30564783 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30564783 Supplier R&D Systems™ Supplier No. LUHM000
  • ONLINE EXCLUSIVE PRICE
    561.00 €
    530.25 € / 1 sets
    Save 30.75 €
    5% Off
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target General Laboratory Research Reagent
Bead Type Magnetic
Kit Contents
  • Standard Cocktail(s)
  • Bead Diluent
  • Biotin Antibody Diluent
  • Calibrator Diluent(s)
  • Streptavidin-PE
  • Wash Buffer Concentrate
  • Mixing Bottles
  • Round-bottom Microplate
  • Foil Plate Sealers
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.