Learn More
Abnova™ Human MASK Partial ORF (NP_057626, 331 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_057626 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51765 |
Molecular Weight (g/mol) | 35.09kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055525
|
Abnova™
H00051765-Q02.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045525
|
Abnova™
H00051765-Q02.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The product of this gene is a member of the GCK group III family of kinases, which are a subset of the Ste20-like kinases. The encoded protein contains an amino-terminal kinase domain, and a carboxy-terminal regulatory domain that mediates homodimerization. The protein kinase localizes to the Golgi apparatus and is specifically activated by binding to the Golgi matrix protein GM130. It is also cleaved by caspase-3 in vitro, and may function in the apoptotic pathway. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: KKPDPKKVQNGAEQDLVQTLSCLSMIITPAFAELKQQDENNASRNQAIEELEKSIAVAEAACPGITDKMVKKLIEKFQKCSADESSpecifications
NP_057626 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.09kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MASK/MST4 | |
RP6-213H19.1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
51765 | |
MASK (Human) Recombinant Protein (Q02) | |
KKPDPKKVQNGAEQDLVQTLSCLSMIITPAFAELKQQDENNASRNQAIEELEKSIAVAEAACPGITDKMVKKLIEKFQKCSADES | |
RUO | |
RP6-213H19.1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |