missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human MCP-1 ELISA Kit

Product Code. 16821506 Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 16821506

Brand: Proteintech™ KE0009196T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous MCP-1

  • Recognizes human MCP-1
  • MCP-1 (Monocyte chemotactic protein 1, CCL2) is a chemokine that can be expressed in monocytes, macrophages and endothelial cells, belongs to CC subfamily of chemokines
    • Chemokines are superfamily of secreted proteins involved in immunoregulatory and inflammatory processes
    • Expression of MCP-1 increases in serum of patients with acute myocarditis
    • Up-regulated in types of CNS injury (ischemia, hemorrhage, trauma, infection, hypoxia, peripheral nerve axotomy)
    • Implicated in pathogenesis of diseases characterized by monocytic infiltrates (psoriasis, rheumatoid arthritis, atherosclerosis)

Process:

  • Monoclonal antibody specific for MCP-1 pre-coated into microwells
  • MCP-1 protein in samples captured by the coated antibody after incubation
  • After extensive washing, monoclonal antibody of biotinylated specific for MCP-1 is added to detect captured MCP-1 protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number NP_002973
Assay Range 31.25 to 1000 pg/mL
Assay Sensitivity 6.9 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 6347
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P13500
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human MMP-9 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.