missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Mesenchymal Stem Cell qPCR Array

Product Code. 17179291
Encompass_Preferred
Change view
Click to view available options
Format:
96-well Plate Type 1
96-well Plate Type 3
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
17179291 96-well Plate Type 1 1 set
17189291 96-well Plate Type 3 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17179291 Supplier STEMCELL Technologies Supplier No. 07541
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For characterization of human MSCs and their differentiation to chondrogenic, adipogenic, or osteogenic lineages

  • The Human Mesenchymal Stem Cell Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of human mesenchymal stem and progenitor cells (MSCs) and their differentiated chondrogenic, adipogenic, or osteogenic progeny
  • MSCs can be expanded in vitro to generate mesenchymal stromal cell cultures, which, under appropriate conditions, can differentiate into adipocytes, chondrocytes, and osteoblasts
  • This qPCR array provides the gene expression profile of MSCs and their trilineage derivatives following in vitro differentiation

Specifications

Description Adipocytes; Chondrocytes; Mesenchymal Stem and Progenitor Cells; Osteoblasts
Format 96-well Plate Type 1
For Use With (Application) Real Time PCR (qPCR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.