missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Mesenchymal Stem Cell Verification Flow Kit

Product Code. 18115061
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
18115061 Store the unopened product at 2°C to 8°C. Do not use past expiration date. 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18115061 Supplier R&D Systems™ Supplier No. FMC020
  • ONLINE EXCLUSIVE PRICE
    1918.32 €
    1704.15 € / 50 tests
    Save 214.17 €
    11% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides single-step staining for the verification of human MSCs

Mesenchymal stromal cells are a heterogeneous population of cultured cells that are adherent to plastic, have a distinct morphology, and express a specific set of marker proteins. Within this heterogeneous population are mesenchymal stem cells. Mesenchymal stem cells are multipotent, self-renewing cells that have the ability to differentiate into adipocytes, chondrocytes, and osteoblasts when cultured in vitro.

Antibodies for MSC verification according to the markers proposed by the International Society for Cellular Therapy
Includes all of the antibodies needed to fulfill the ISCT minimal criteria for the definition of human MSCs
Can be used with the Human MSC Functional ID kit to define MSCs according to the ISCT recommendations
Provides a cost-effective method to analyze several MSC markers in a single sample
Reveals heterogeneity in the starting cell population

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.