missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MINA53 Synthetic Peptide

Product Code. 15295019
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15295019 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15295019 Supplier Invitrogen™ Supplier No. PEP1023
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 15 amino acids near the amino terminus of human MINA. PEP-1023 can be used as a blocking peptide with polyclonal antibody PA5-20909.

MINA is nuclear localized, myc-inducible protein that is thought to play a role in mammalian cell proliferation. Treatment of cancer cells lines such as the colon cancer cell line SW680 with siRNA against MINA inhibits cell growth, demonstrating that MINA may be a potential therapeutic target. MINA regulates several genes related to cell adhesion and metabolism that have also been shown to be regulated by c-Myc, but also regulates other genes whose expression are not modulated by c-Myc such as EGFR, IL-6 and HGF. MINA has also been found to act as a repressor to IL-4 expression in T cells, indicating that it may also play a role in T cell differentiation and genetic variation in T helper type 2 bias.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 84864
Species Human
Purification Method purified
Accession Number Q8IUF8
Common Name MINA53
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1810047J07Rik; 2410057H13Rik; 3830408E23Rik; 60S ribosomal protein L27a histidine hydroxylase; AI449204; Bifunctional lysine-specific demethylase and histidyl-hydroxylase MINA; DKFZp762O1912; FLJ14393; histone lysine demethylase MINA; MDIG; mina; mina {ECO:0000312; Mina53; Mina-53; mineral dust induced gene protein; mineral dust-induced gene protein; MYC induced nuclear antigen; myc-induced nuclear antigen; myc-induced nuclear antigen, 53 kDa; NO52; Nucleolar protein 52; Protein mina53; RGD:708521}; Ribosomal oxygenase 2; Ribosomal oxygenase MINA; RIOX2; ROX
Gene Symbol RIOX2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.