missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat Activin A Protein

Product Code. p-7112703
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Unit Size:
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18760428 Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 500µg
18740169 Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18760428 Supplier R&D Systems™ Supplier No. 338AC500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHO derived

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08476
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 3624
Molecular Weight (g/mol) M.W. (Observed): 14 kDa, reducing conditions 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer)
Quantity 500 μg
Source Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol INHBA
Biological Activity 0.2 to 1.2ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human, Mouse, Rat
Recombinant Recombinant
Reconstitution Reconstitute at 100-500 μg/mL in sterile 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein Activin A
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.