missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human/Mouse SMN ELISA Kit

Product Code. 16831456 Shop All Proteintech Products
Click to view available options
Target Species:
Human, Mouse
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 16831456

Brand: Proteintech™ KE0002796T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KE00027 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The SMN ELISA kit is designed for the quantitative determination of SMN in samples of human and mouse origin

  • An antibody specific for SMN has been pre-coated onto the microwells.
  • The SMN protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody specific for SMN is added to detect the captured SMN protein.
  • For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
  • Survival Motor Neuron (SMN) protein required for efficient assembly of small nuclear ribonucleoprotein (snRNP) complexes is encoded by nearly identical telomeric and centromeric forms of SMN gene respectively.
  • Both the SMN1 and SMN2 genes express SMN protein, however, the amount of functional full-length protein produced by SMN2 is much less than that produced by SMN1 due to the alternative splicing.
  • The SMN gene is constitutively expressed in wide variety of tissues including brain, kidney, liver and spinal cord, while motor neurons are particularly vulnerable to reduced SMN protein levels. Deletion or mutational inactivation of the SMN1 gene causes spinal muscular atrophy (SMA), a lethal genetic disease characterized by loss of motor neurons in the spinal cord.
  • The absence of SMN1 can be partially compensated for by SMN2 and the SMN2 expression level is associated with SMA severity. This kit is for the quantitative determination of SMN protein level in vivo.

Specifications

Accession Number NP_059107
Assay Range 62.5 to 4000 pg/mL
Assay Sensitivity 6 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human, Mouse
Gene ID (Entrez) 6607
Host Species Human/Mouse
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID Q16637
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human B7-H3/CD276 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.