missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MYO6 Partial ORF (NP_004990.2, 1188 a.a. - 1285 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
Description
This gene encodes a protein involved intracellular vesicle and organelle transport, especially in the hair cell of the inner ear. Mutations in this gene have been found in patients with non-syndromic autosomal dominant and recessive hearing loss. [provided by RefSeq]
Sequence: KKKGWWYAHFDGPWIARQMELHPDKPPILLVAGKDDMEMCELNLEETGLTRKRGAEILPRQFEEIWERCGGIQYLQNAIESRQARPTYATAMLQSLLK
Specifications
Specifications
| Accession Number | NP_004990.2 |
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 4646 |
| Molecular Weight (g/mol) | 36.52kDa |
| Name | MYO6 (Human) Recombinant Protein (Q01) |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 ug |
| Immunogen | KKKGWWYAHFDGPWIARQMELHPDKPPILLVAGKDDMEMCELNLEETGLTRKRGAEILPRQFEEIWERCGGIQYLQNAIESRQARPTYATAMLQSLLK |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction