missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NBAS (aa 962-1047) Control Fragment Recombinant Protein

Product Code. 30198979
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30198979 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30198979 Supplier Invitrogen™ Supplier No. RP96139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57831 (PA5-57831. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Negatively regulates nitric oxide production by inducing NOS1 and NOS3 translocation to actin cytoskeleton and inhibiting their enzymatic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A2RRP1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51594
Name Human NBAS (aa 962-1047) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4933425L03Rik; ILFS2; NAG; NAG/BC035112 fusion; NAG/FAM49A fusion; NBAS; neuroblastoma amplified sequence; neuroblastoma-amplified gene protein; neuroblastoma-amplified protein; neuroblastoma-amplified protein-like; neuroblastoma-amplified sequence; similar to neuroblastoma-amplified protein; SOPH
Common Name NBAS
Gene Symbol NBAS
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AKGDLKFPLKIFQHSKPDLQQKIIPDQDQLMAIALECIYTCERNDQLCLCYDLLECLPERGYGDKTEATTKLHDMVDQLEQILSVS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.