missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NEFL ProQuantum Immunoassay Kit
Click to view available options
Quantity:
5 X 96 Tests
Unit Size:
Each
Description
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220 kDa, with some variation in different species.
Specifications
Specifications
| Accession Number | P07196, P08551, P19527 |
| Assay Range | 0.48 to 7500 pg/mL |
| Assay Sensitivity | 0.48 pg/mL |
| Conjugate | FAM™ |
| Product Type | ProQuantum |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | qPCR Instruments |
| Gene ID (Entrez) | 18039, 4747, 83613 |
| Gene Symbol | NEFL |
| Kit Contents | Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction