Learn More
Abcam Human NEIL1 knockout HEK-293T cell line
Product Code. 32157606 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32157606 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32157606 Supplier Abcam Supplier No. ab2661672x1000000Cells_
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
NEIL1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select '...
NEIL1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells