missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NF-L ELISA Kit

Product Code. 30535588
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30535588 96 Tests Pack of 96
30534577 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30535588 Supplier Invitrogen™ Supplier No. BMS2362
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ELISA

The Human NF-L ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of human NF-L in CSF, serum, and plasma (citrate and heparin) and cell culture supernatants (CCS).

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220 kDa, with some variation in different species.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07196, P08551, P19527
Assay Range 1.6-1200
Assay Sensitivity 0.8 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Cerebrospinal Fluid, Plasma, Serum, Supernatant
Target Species Human, Mouse, Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 68 kDa neurofilament protein; AI847934; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; micro glutamic acid-rich protein; NEF3; NEFL; nefl.L; neurofilament; neurofilament 3; neurofilament L subunit; neurofilament light; neurofilament light polypeptide; neurofilament protein; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet L protein; neurofilament, light L homeolog; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament, light polypeptide L homeolog; neurofilament-l; neurofilament-light; Nf68; NFL; N-FL; NF-L; NFM; NF-M; NLC; OTTHUMP00000196614; PPP1R110; protein phosphatase 1, regulatory subunit 110; XELAEV_18018673mg; XNF-L68 kDa neurofilament protein; AI847934; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; micro glutamic acid-rich protein; NEF3; NEFL; nefl.L; neurofilament; neurofilament 3; neurofilament L subunit; neurofilament light; neurofilament light polypeptide; neurofilament protein; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet L protein; neurofilament, light L homeolog; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament, light polypeptide L homeolog; neurofilament-l; neurofilament-light; Nf68; NFL; N-FL; NF-L; NFM; NF-M; NLC; OTTHUMP00000196614; PPP1R110; protein phosphatase 1, regulatory subunit 110; XELAEV_18018673mg; XNF-L;
Gene ID (Entrez) 18039, 4747, 83613
Gene Symbol NEFL
Interassay CV 4%
Intraassay CV 3.7%
Kit Contents Pre-coated microwell plate, Standard, Biotinylated Detection Antibody, Streptavidin-HRP, Sample Diluent, Wash Buffer Concentrate, Assay Buffer concentrate, Stop Solution, Substrate Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target NF-L
Test Time 1 hr 20 min
Total Assay Time 3 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.