missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human NGAL ELISA Kit

Product Code. 16811556 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species unitSize
16811556 Human 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16811556 Supplier Proteintech™ Supplier No. KE0014496T
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous NGAL

  • Recognizes human NGAL
  • NGAL (Neutrophil gelatinase-associated lipocalin, Lipocalin-2, LCN2) is a secreted glycoprotein that belongs to lipocalin protein family
    • NGAL exists as a monomer, homodimer, disulfide-linked heterodimer in cells, covalently conjugated with gelatinase in order to decrease degradation
    • Roles include iron trafficking, chemotactic, promote cell differentiation and proliferation
    • Found in human neutrophils and expressed in tissues containing renal, uterus, liver and spleen, cardiomyocytes, neurons, immune cells
    • NGAL is a biomarker of cardiovascular diseases, ischemic stroke, acute renal injury, inflammation, metabolic diseases
    • Detected in human serum, saliva, urine

Process:

  • Antibody specific for NGAL pre-coated onto microwells
  • NGAL protein in samples captured by the coated antibody after incubation
  • After extensive washing, another horseradish peroxidase (HRP)-conjugated antibody specific for NGAL is added to detect captured NGAL protein
  • For signal development, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number NM_005564
Assay Range 78.1 to 5000 pg/mL
Assay Sensitivity 3.3 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 3934
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID P80188
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human CD147 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.