missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NINJ1 Partial ORF (NP_004139.1, 1 a.a. - 66 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Spécification
Accession Number | NP_004139.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4814 |
Molecular Weight (g/mol) | 33kDa |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
16044465
|
Abnova™
H00004814-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054465
|
Abnova™
H00004814-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The ninjurin protein is upregulated after nerve injury both in dorsal root ganglion neurons and in Schwann cells (Araki and Milbrandt, 1996 [PubMed 8780658]). It demonstrates properties of a homophilic adhesion molecule and promotes neurite outgrowth from primary cultured dorsal root ganglion neurons.[supplied by OMIM]
Sequence: MDSGTEEYELNGGLPPGTPGSPDASPARWGWRHGPINVNHYASKKSAAESMLDIALLMANASQLKASpécification
NP_004139.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NIN1/NINJURIN | |
NINJ1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4814 | |
NINJ1 (Human) Recombinant Protein (Q01) | |
MDSGTEEYELNGGLPPGTPGSPDASPARWGWRHGPINVNHYASKKSAAESMLDIALLMANASQLKA | |
RUO | |
NINJ1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |