missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Noggin Recombinant Protein, PeproTech®
GREENER_CHOICE

Product Code. 17850123
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17850123 50 μg 50µg
17840123 100 μg 100µg
17880143 2 x 500 μg 1mg
30087766 2 x 500 μg Pack of 2
17811063 20 μg 20µg
17830873 250 μg 250µg
17850883 500 μg 500µg
17861183 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17850123 Supplier Gibco™ Supplier No. 12010C50UG

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Recombinant Protein

120-10C-1MG will be provided as 2 x 500 μg (120-10C-500UG). Recombinant Human Noggin is a 46 kDa disulfide-linked homodimer consisting of two 205 amino acid polypeptide chains. Monomeric glycosylated noggin migrates at an apparent molecular weight of approximately 28.0-33.0 kDa by SDS PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13253
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 9241
Molecular Weight (g/mol) 46 kDa
Name Human Noggin
Purification Method Purified
Quantity 50 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A
Common Name Noggin
Gene Symbol NOG
Biological Activity Determined by its ability to inhibit 5.0 ng/ml of BMP-4 induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The expected ED50 for this effect is 2.0-3.0 ng/ml of Noggin.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence QHYLHIRPAP SDNLPLVDLI EHPDPIFDPK EKDLNETLLR SLLGGHYDPG FMATSPPEDR PGGGGGAAGG AEDLAELDQL LRQRPSGAMP SEIKGLEFSE GLAQGKKQRL SKKLRRKLQM WLWSQTFCPV LYAWNDLGSR FWPRYVKVGS CFSKRSCSVP EGMVCKPSKS VHLTVLRWRC QRRGGQRCGW IPIQYPIISE CKCSC
Content And Storage -20°C
Activity Determined by its ability to inhibit 5.0 ng/ml of BMP-4 induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The expected ED50 for this effect is 2.0-3.0 ng/ml of Noggin.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.