missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Notch-1 (NP_060087) VersaClone cDNA
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
missing translation for 'unitSize'
10µg
Beschreibung
The Human Notch-1 (NP_060087) VersaClone cDNA clone contains the complete ORF for Notch-1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Notch-1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spezifikation
Spezifikation
| Gene | hNOTCH1 |
| Accession Number | NP_060087 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl |
| Insert Type | 7682bp |
| pH | 8.5 |
| Gene ID (Entrez) | 4851 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur