missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Notch-2 (NP_077719.2) VersaClone cDNA

Product Code. 18781967
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18781967 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18781967 Supplier R&D Systems™ Supplier No. RDC3540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene Notch-2
Accession Number NP_077719.2
Gene ID (Entrez) 4853
Gene Alias AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2 Notch homolog 2 (Drosophila), Notch homolog 2, Notch2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.