Learn More
Abcam Human NSDHL knockout HEK-293T cell line
Product Code. 32140355 Shop All Abcam Products
Change view
Click to view available options
2x1000000 Cells
Product Code.
32140355 2x1000000 Cells
1 options
Product Code. 32140355 Supplier Abcam Supplier No. ab2666822x1000000Cells_
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
NSDHL KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp deletion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout...
NSDHL KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp deletion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells