missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human NTCP (G) SLC Transporter Cells

Product Code. 17146632 Shop All Gibco Products
Change view
Click to view available options
Quantity:
8–10M cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17146632 8–10M cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17146632 Supplier Gibco™ Supplier No. GM1113G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HEK293 cells that transiently overexpress the NTCP solute carrier (SLC) transport protein

  • TRANSiPORT Human NTCP (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the NTCP solute carrier (SLC) transport protein. Solute carriers are a super-family of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes in and out of cells. The NTCP SLC transporter protein is found in hepatocytes.
  • Assess potential for transporter-mediated drug metabolism
  • Easy to use format
  • Obtain high quality results with a large signal-to-noise ratio
  • Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
  • Mock HEK293 cells (Cat. No. GM1101G) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of transport due to the presence of endogenous transporters or non-specific binding.
  • The convenient product format enables data generation in 2 days from thawing of the cells to final results
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen vapor phase.
Cell Line 293 (HEK)
Culture Type Mammalian Cell Culture
Species Human
No. of Cells 8-10 x 106
Product Type SLC Transporter Cell
Quantity 8–10M cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.