missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ QuicKit Immunoassay Control Set 1209 for Human Perforin
Click to view available options
:
1 Each
Specifications
Specifications
| Target Species | Human |
| Gene ID (Entrez) | 5551 |
| Quantity | 1 Each |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFP, PFPcytolysin, PRF1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction