missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PABPC1L2A (aa 165-200) Control Fragment Recombinant Protein

Product Code. 30199585
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199585

Brand: Invitrogen™ RP104326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59629 (PA5-59629. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Tekniske data

Accession Number Q5JQF8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 340529
Name Human PABPC1L2A (aa 165-200) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias PABPC1L2; Pabpc1l2a; PABPC1L2B; poly(A) binding protein cytoplasmic 1 like 2 A; poly(A) binding protein, cytoplasmic 1-like 2 A; poly(A) binding protein, cytoplasmic 1-like 2 B; polyadenylate-binding protein 1-like 2; RBM32A; RBM32B; RGD1566367; RNA binding motif protein 32 A; RNA binding motif protein 32 B; RNA-binding motif protein 32; RNA-binding protein 32
Common Name PABPC1L2A
Gene Symbol PABPC1L2A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KSHKEREAERGAWARQSTSADVKDFEEDTDEEATLR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.