missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PAM16 (aa 73-125) Control Fragment Recombinant Protein

Product Code. 30199026
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30199026 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30199026 Supplier Invitrogen™ Supplier No. RP105641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66523 (PA5-66523. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Regulates ATP-dependent protein translocation into the mitochondrial matrix. Inhibits DNAJC19 stimulation of HSPA9/Mortalin ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y3D7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51025
Name Human PAM16 (aa 73-125) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2010110I09Rik; AV006767; CGI-136; MAGMAS; magmas processed gene 1; magmas-like protein; mitochondria associated granulocyte macrophage CSF signaling molecule; mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction; mitochondria-associated granulocyte macrophage CSF-signaling molecule; mitochondria-associated protein involved in granulocyte-macrophage colony-stimulating factor signal transduction; mitochondrial import inner membrane translocase subunit TIM16; Pam16; presequence translocase-associated motor 16 homolog (S. cerevisiae); presequence translocase-asssociated motor 16 homolog (S. cerevisiae); presequence translocated-associated motor subunit PAM16; SMDMDM; TIM16; Timm16
Common Name PAM16
Gene Symbol PAM16
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VQKNYEHLFKVNDKSVGGSFYLQSKVVRAKERLDEELKIQAQEDREKGQMPHT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.