missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PCPE-1 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Tamaño de la unidad:
Each
Pack of 96
Descripción
Human PCPE-1 quantitates human PCPE-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PCPE-1.
Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity.
Especificaciones
Especificaciones
| Accession Number | astroglial cell transcript 2; Astt2; Astt-2; P14; Pcolce; PCPE; PCPE1; PCPE-1; procollagen C-endopeptidase enhancer; procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen COOH-terminal proteinase enhancer 1; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; procollagen, type 1, COOH-terminal proteinase enhancer; Type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancer |
| Assay Range | 51 to 12,500 pg/mL |
| Assay Sensitivity | 51 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 5118 |
| Gene Symbol | PCOLCE |
| Interassay CV | <12% |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido