Learn More
Abcam Human PHF23 knockout HEK-293T cell line
Product Code. 32142791 Shop All Abcam Products
Change view
Click to view available options
2x1000000 Cells
Product Code.
32142791 2x1000000 Cells
1 options
Product Code. 32142791 Supplier Abcam Supplier No. ab2666242x1000000Cells_
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PHF23 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: Insertion of the selection cassette in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only kn...
PHF23 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: Insertion of the selection cassette in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells