missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-VEGFR2/KDR DuoSet IC ELISA
Description
The Human Phospho-VEGFR2/KDR DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human VEGFR2/KDR/Flk-1.
Specifications
Specifications
| Product Type | ELISA Development Kit for Phosphorylated Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3791 |
| Synonym | CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2 |
| Packaging Quantity | 15 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?