missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human PLA2G3 (NP_056530.2) VersaClone cDNA
Specifications
Specifications
| Gene | PLA2G3 |
| Accession Number | NP_056530.2 |
| Concentration | 10 μg (at 0.2 μg/μL) |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1543 bp |
| Gene ID (Entrez) | 50487 |
| Gene Alias | EC 3.1.1.4, GIII SPLA2, GIII-SPLA2, group 3 secretory phospholipase A2, group III secreted phospholipase A2, Group III secretory phospholipase A2, Phosphatidylcholine 2-acylhydrolase 3, phosphatidylcholine 2-acylhydrolase GIII, phospholipase A2, group III, sPLA2-III, SPLA2III |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?