missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLAC2 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1159
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PLAC2 Synthetic Peptide, PEP-1159, from Invitrogen.
This peptide corresponds to 17 amino acids near the center of human PLAC2.PEP-1159 can be used as a blocking peptide with polyclonal antibody PA5-21045.Specifications
257000 | |
purified | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TINCR | |
0.02% sodium azide | |
50 μg | |
PBS |
Human | |
Human PLAC2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |