Learn More
Abnova™ Human PLAU (high molecular weight) native Protein from human urine
Description
This gene encodes a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. A specific polymorphism in this gene may be associated with late-onset Alzheimer's disease and also with decreased affinity for fibrin-binding. This protein converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. Plasmin in turn cleaves this protein at a Lys-Ile bond to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
| Concentration | 3.27 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | In 0.05M sodium acetate, 0.1M NaCl, pH 5.0 (1mM EDTA) |
| Gene ID (Entrez) | 5328 |
| Molecular Weight (g/mol) | 54kDa |
| Name | PLAU (Human) Native Protein |
| Preparation Method | Native protein purified from human urine |
| Purification Method | Column chromatography using several resins |
| Quantity | 100 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.