missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Plexin A2 (NP_079455) VersaClone cDNA

Product Code. 18746919
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18746919 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18746919 Supplier R&D Systems™ Supplier No. RDC1208
  • ONLINE EXCLUSIVE PRICE
    1240.00 €
    1170.75 € / 10µg
    Save 69.25 €
    5% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Tilføj til indkøbskurv
Tilføj til indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

It participates in cell migration and axon guidance, and does so by serving as a receptor for Sema6A and B, and as a coreceptor with neuropilin 1 for Sema3A and C

The Human Plexin A2 (NP_079455) VersaClone cDNA clone contains the complete ORF for Plexin A2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Plexin A2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Ordreinfo

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Tekniske data

Accession Number NP_079455
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 5697bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.