missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Pluripotent Stem Cell Naïve State qPCR Array

Product Code. 17129291
Encompass_Preferred
Change view
Click to view available options
Format:
96-well Plate Type 3
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
17129291 96-well Plate Type 3 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17129291 Supplier STEMCELL Technologies Supplier No. 07523

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For characterization of gene expression associated with hPSC naïve or primed states

  • Characterize your hPSCs by accurately measuring their gene expression profile to determine their status along the spectrum from naïve to primed pluripotency
  • The Human Pluripotent Stem Cell (hPSC) Naïve State qPCR Array has validated primers and probes to detect 90 genes preselected for their demonstrated differential expression in naïve state and primed hPSCs and their derivatives, as well as 6 endogenous control genes
  • TBP (TATA box-binding protein) qPCR Control Template is provided as a synthetic DNA positive control
  • Naïve state hPSCs are self-renewing, retain the properties of the pre-implantation blastocyst, and can differentiate to both embryonic and extra-embryonic lineages
  • Primed hPSCs are also self-renewing, but they are restricted to differentiation along the three embryonic germ layers

Specifications

Description Pluripotent Stem Cells
Format 96-well Plate Type 3
For Use With (Application) Real Time PCR (qPCR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.