missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PMCA4 ATPase (aa 1168-1241) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (53%), Rat (53%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83500 (PA5-83500. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin (Cat. # MA3-917 and MA3-918).
Specifications
Specifications
| Accession Number | P23634 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 493 |
| Name | Human PMCA4 ATPase (aa 1168-1241) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | ATP2B2; ATP2B4; ATPase plasma membrane Ca2+ transporting 4; ATPase, Ca++ transporting, plasma membrane 4; ATPase, Ca++ transporting, plasma membrane 4 ATP synthase, H+ transporting, mitochondrial F1; ATPase, Ca++ transporting, plasma membrane 4 tv2; matrix-remodeling-associated protein 1; MXRA1; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; plasma membrane calcium ATPase 4; plasma membrane calcium ATPase 4 variant b; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium pump; Plasma membrane calcium pump isoform 4; plasma membrane calcium-pumping ATP-ase; plasma membrane calcium-transporting ATPase 4; PMCA4; PMCA4b; PMCA4x; sarcolemmal calcium pump |
| Common Name | PMCA4 ATPase |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction