missing translation for 'onlineSavingsMsg'
Learn More

Abcam Human Podoplanin ELISA Kit- Intracellular

Product Code. 32115605 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 384 Tests
1 x 96 Tests
10 x 96 Tests
Unit Size:
384 tests
96 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115605 10 x 96 Tests 96 tests
32115311 1 x 96 Tests 96 tests
32115189 1 x 384 Tests 384 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 32115605 Fournisseur Abcam Code fournisseur ab32392410x96Tests
 Restricted Item
Estimated Shipment: 26-11-2026
Ajouter au panier
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Podoplanin ELISA Kit- Intracellular

Human Podoplanin ELISA Kit- Intracellular

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Accession Number Q86YL7
Assay Range 25 to 1600 pg/mL
Assay Sensitivity 7 pg/mL
Content And Storage 4°C
Detectable Analytes PDPN (Podoplanin)
Assay Human Podoplanin ELISA Kit- Intracellular
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Sample Type Cell Lysates
Sufficient For 10 x 96 Tests
For Use With (Application) ELISA
Gene PDPN
Immunoassay Kit Grade RUO
Measuring Range 25 to 1600 pg/mL
Protein PDPN
Quantitation Range 25 to 1600 pg/mL
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Immuno-oncology
Shipping Condition Blue Ice
Target PDPN (Podoplanin)
Target Kit Named PDPN (Podoplanin)
UniProt ID Q86YL7
Detection Limit 25 pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.