Learn More
Abcam Human PPID (Cyclophilin 40) knockout HeLa cell line
Product Code. 32140268 Shop All Abcam Products
Change view
Click to view available options
2x1000000 Cells
Product Code.
32140268 2x1000000 Cells
1 options
Product Code. 32140268 Supplier Abcam Supplier No. ab2657882x1000000Cells_
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PPID KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout...
PPID KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells