missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Progesterone Receptor (aa 533-547) Synthetic Peptide

Product Code. 11594670
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 11594670

Brand: Invitrogen™ PEP016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Progesterone Receptor (PR) (residues 533-547) Synthetic Peptide, PEP-016, from Invitrogen.

The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities. PEP-016 is a 15 amino acid (amino acids 533-547) synthetic peptide whose sequence is found N-terminal to the PR DNA binding domain. The sequence of this peptide is (amino to carboxy terminus): G-L-P- Q-V-Y-P-P-Y-L-N-Y-L-R-PThis peptide may be used for neutralization and control experiments with the monoclonal antibody that reacts with this product and PR, catalog &Num; MA1-410. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 5241
Species Human
Purification Method purified
Accession Number P06401
Common Name Human Progesterone Receptor (aa 533-547)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol PGR
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence G - L - P- Q - V - Y -P - P -Y -L - N - Y - L - R - P
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.