missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Prolactin R (NP_000940) VersaClone cDNA
Description
- PRLR prolactin receptor [ Homo sapiens (human) ]
- Also known as: HPRL; MFAB; hPRLrI; RI-PRLR
- The Prolactin R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- PRLR is a receptor for the anterior pituitary hormone, prolactin, and belongs to the type I cytokine receptor family.
- Prolactin-dependent signaling occurs as the result of ligandinduced dimerization of the prolactin receptor.
- Several alternatively spliced transcript variants encoding different membrane-bound and soluble isoforms have been described for PRLR, which may function to modulate the endocrine and autocrine effects of prolactin in normal tissue and cancer.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hPRLR |
| Accession Number | NP_000940 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1882 bp |
| Gene ID (Entrez) | 5618 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?