missing translation for 'onlineSavingsMsg'
Learn More

Human Promyelocytic Leukemia (HL-60) Total RNA for QuantiGene Assays

Product Code. 32096700
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32096700 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32096700 Supplier Invitrogen Supplier No. QS0607
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Cell Type HL-60 Cell
Species Human
Sample Type RNA
For Use With (Application) Nucleic Acid Quantitation
Product Type Total RNA
Quantity 10 μg
Sample Source Tumor or Diseased Tissue, Cell Line
Shipping Condition Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.