missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRR5L Synthetic Peptide

Product Code. 15205249
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15205249 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15205249 Supplier Invitrogen™ Supplier No. PEP1243
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 17 amino acids near the center of human PRR5L. PEP-1243 can be used as a blocking peptide with polyclonal antibody PA5-21129.

Proline-rich protein 5-like (PRR5L), also known as Protor-2, is structurally similar to PRR5 and also binds to the mTOR complex 2 (mTORC2). PRR5L is not required for mTORC2 integrity or kinase activity, but dissociates from mTORC2 upon knockdown of tuberous sclerosis complex 1 (TSC1) and TSC2. Knockdown of PRR5L reduced apoptosis in mTORC2 deficient cells as well as cells where mTOR was active, suggesting that mTORC2-dissociated PRR5L may promote apoptosis, and thus play a role in the balance between cell growth and cell death.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 79899
Species Human
Purification Method purified
Accession Number Q6MZQ0
Common Name PRR5L
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 2600010E01Rik; 4833411O04Rik; proline rich 5 like; proline-rich protein 5-like; Protein observed with Rictor-2; PROTOR2; protor-2; Prr5l
Gene Symbol PRR5L
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.