missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRRT2 Synthetic Peptide

Product Code. 15235509
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15235509 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15235509 Supplier Invitrogen™ Supplier No. PEP1580
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 18 amino acids near the center of human PRRT2. PEP-1580 can be used as a blocking peptide with polyclonal antibody PA5-34537. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37°C.

The proline-rich transmembrane protein 2 (PRRT2) contains a proline-rich domain in its N-terminal half and is predominantly expressed in brain and spinal cord in embryonic and postnatal stages. While little is known of the function of this protein, mutations in PRRT2 have been shown to be the causative gene of paroxysmal kinesigenic dyskinesia, which is characterized by recurrent, brief attacks of abnormal involuntary movements induced by sudden voluntary movements. Recent studies have shown that PRRT2 may also be involved in some forms of benign familial infantile epilepsy (BFIE), an autosomal dominant epilepsy syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 112476
Species Human
Purification Method purified
Accession Number Q7Z6L0
Common Name PRRT2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1500031I19Rik; AI195361; BFIC2; BFIS2; Dispanin subfamily B member 3; DSPB3; dystonia 10; DYT10; EKD1; FICCA; ICCA; IFITMD1; infantile convulsions and paroxysmal choreoathetosis; interferon induced transmembrane protein domain containing 1; PKC; proline rich transmembrane protein 2; proline-rich transmembrane protein 2; PRRT2; RGD1564195
Gene Symbol PRRT2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.