missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PTPN13 (aa 1187-1292) Control Fragment Recombinant Protein

Product Code. 30199528
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199528

Brand: Invitrogen™ RP108381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139860 (PA5-139860. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q12923
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5783
Name Human PTPN13 (aa 1187-1292) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI324989; DKFZp686J1497; FAP-1; Fas-associated protein-tyrosine phosphatase 1; hPTP1E; hPTPE1; PNP1; protein Tyr phosphatase; protein tyrosine phosphatase DPZPTP; Protein tyrosine phosphatase PTP-BL; protein tyrosine phosphatase, non-receptor type 13; protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associated phosphatase); protein-tyrosine phosphatase 1 E; Protein-tyrosine phosphatase PTPL1; protein-tyrosine phosphatase RIP; ptp; Ptp14; PTP1E; PTP36; PTP-BAS; PTP-BL; PTP-E1; PTPL1; PTPLE; PTPN13; Ptpri; RIP; tyrosine phosphatase; tyrosine-protein phosphatase non-receptor type 13
Common Name PTPN13
Gene Symbol PTPN13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence STPVHLTNEMKNYMKKSSYMQDSAIDSSSKDHHWSRGTLRHISENSFGPSGGLREGSLSSQDSRTESASLSQSQVNGFFASHLGDQTWQESQHGSPSPSVISKATE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.