missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human QDPR (aa 36-125) Control Fragment Recombinant Protein

Product Code. 30210484
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210484

Brand: Invitrogen™ RP103971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64233 (PA5-64233. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the enzyme dihydropteridine reductase, which catalyzes the NADH-mediated reduction of quinonoid dihydrobiopterin. This enzyme is an essential component of the pterin-dependent aromatic amino acid hydroxylating systems. Mutations in this gene resulting in QDPR deficiency include aberrant splicing, amino acid substitutions, insertions, or premature terminations. Dihydropteridine reductase deficiency presents as atypical phenylketonuria due to insufficient production of biopterin, a cofactor for phenylalanine hydroxylase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09417
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5860
Name Human QDPR (aa 36-125) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610008L04Rik; 6,7-dihydropteridine reductase; D5Ertd371e; Dhpr; dihydropteridine reductase; HDHPR; HPR; PKU2; QDPR; Quinoid dihydropteridine reductase; SDR33C1; Short chain dehydrogenase/reductase family 33 C member 1; short chain dehydrogenase/reductase family 33 C, member 1; testis secretory sperm-binding protein Li 236 P
Common Name QDPR
Gene Symbol QDPR
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence WVASVDVVENEEASASIIVKMTDSFTEQADQVTAEVGKLLGEEKVDAILCVAGGWAGGNAKSKSLFKNCDLMWKQSIWTSTISSHLATKH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.