Learn More
ACROBiosystems Human R-Spondin 1 / RSPO1 Protein, His Tag
Description
R-spondin-1 is also known as Roof plate-specific Spondin 1 (RSPO1) and cysteinerich and single thrombospondin domain containing protein 3 (Cristin 3), which is a secreted protein which belongs to the R-Spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. All Rspondins regulate Wnt/β-catenin signaling, but have distinct expression patterns. Like other R-Spondins, R-Spondin-1 contains two adjacent cysteinerich furinlike domains (aa 34-135) with one potential N-glycosylation site, followed by a thrombospondin (TSP1) motif (aa 147-207) and a region rich in basic residues (aa 211-263). Only the furinlike domains are needed for β-catenin stabilization. A putative nuclear localization signal at the C-terminus may allow some expression in the nucleus.
Specifications
Specifications
| Accession Number | NP_001033722.1 |
| Molecular Weight (g/mol) | 28.7 kDa |
| Quantity | 50 μg |
| Source | Human R-Spondin 1, His Tag (RS1-H4221) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 263 (Accession # Q2MKA7-1). |
| Immunogen | Ser 21 - Ala 263 |
| Storage Requirements | -20°C |
| Gene Alias | RSPO1,CRISTIN3,FLJ40906,RP11-566C13.1,R-spondin-1 |
| Common Name | R-Spondin 1 |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.