missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human R-Spondin 3 (NP_116173.2) VersaClone cDNA

Product Code. 18703287
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18703287 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18703287 Supplier R&D Systems™ Supplier No. RDC3211
  • ONLINE EXCLUSIVE PRICE
    348.88 €
    309.75 € / 10µg
    Save 39.13 €
    11% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene R-Spondin 3
Accession Number NP_116173.2
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 832 bp
Gene ID (Entrez) 84870
Gene Alias Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Show More Show Less
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.