missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human RANK Receptor (soluble) Recombinant Protein, PeproTech®

Product Code. p-7246268
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
This item is not returnable. View return policy

Product Code. 17842603

Brand: Gibco™ 31008100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human sRANK Receptor is a 19.3 kDa polypeptide containing the TNFR-homologous, cysteine-rich portion of the extracellular domain of RANK receptor (175 amino acid residues). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6Q6
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 8792
Molecular Weight (g/mol) 19.3 kDa
Name Human RANK Receptor (soluble)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11 A; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11 A; tumor necrosis factor receptor superfamily member 11 A, NFKB activator; tumor necrosis factor receptor superfamily, member 11 A, NFKB activator
Common Name RANK (CD265)
Gene Symbol TNFRSF11A
Biological Activity Determined by its ability to inhibit sRANKL induced NF-kappaB in RAW264.7 cells in the absence of any cross-linking. The expected ED50 for this effect in the presence of 15 ng/ml of recombinant sRANKL, is 30-50 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQIAPPCTSE KHYEHLGRCC NKCEPGKYMS SKCTTTSDSV CLPCGPDEYL DSWNEEDKCL LHKVCDTGKA LVAVVAGNST TPRRCACTAG YHWSQDCECC RRNTECAPGL GAQHPLQLNK DTVCKPCLAG YFSDAFSSTD KCRPWTNCTF LGKRVEHHGT EKSDAVCSSS LPARK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.